Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199187.2 | NP_001186116.1 | 6 | 646 | ||
NM_026937.3 | NP_081213.1 | 6 | 659 | |||
GenBank mRNA | AK007519.1 | - | 6 | 643 | ||
AK139944.1 | - | 6 | 646 | |||
AK171958.1 | - | 6 | 620 | |||
AK179536.1 | - | 2 | 159 | |||
BC030905.1 | - | 6 | 625 | 4503445 |