Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271777.1 | NP_001258706.1 | 4, 5 | 508 | ||
NM_027062.2 | NP_081338.1 | 4, 5 | 729 | |||
GenBank mRNA | AK005962.1 | - | 4, 5 | 512 | ||
BC019967.1 | - | 4, 5 | 729 | 4023907 |