Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027412.3 | NP_081688.2 | 3 | 597 | ||
GenBank mRNA | AK008753.1 | - | 2 | 928 | ||
AK018145.1 | - | 3 | 637 | |||
AK169288.1 | - | 3 | 598 | |||
AK169296.1 | - | 3 | 598 | |||
BC035304.1 | - | 2 | 508 | |||
BC043061.1 | - | 2 | 909 | 5704924 | ||
BC058967.1 | - | 2 | 909 | 5704924 |