Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 1, 2 | 313 | ||
GenBank mRNA | BC063268.1 | - | 1, 2 | 109 | 4187769 | |
BC080688.1 | - | 1, 2 | 255 | 6515742 | ||
CJ171970.1 | - | 1, 2 | 313 |