Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029078.3 | NP_083354.3 | 8 | 2614 | ||
GenBank mRNA | AK133347.1 | - | 4 | 1295 | ||
AK173035.1 | - | 8 | 2586 | |||
BC027361.1 | - | 1 | 107 | |||
BC150781.1 | - | 8 | 2450 | 9087692 | ||
EF537860.1 | - | 5 | 1718 | |||
EF537861.1 | - | 5 | 1718 |