Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037841.3 | NP_001032930.1 | 2 | 346 | ||
NM_001286383.1 | NP_001273312.1 | 2 | 346 | |||
NM_029295.3 | NP_083571.1 | 2 | 346 | |||
GenBank mRNA | AF401530.1 | - | 2 | 251 | ||
AF401531.2 | - | 2 | 251 | |||
AK005577.1 | - | 2 | 306 | |||
AK032759.1 | - | 2 | 276 | |||
AK046397.1 | - | 2 | 298 | |||
AK050112.1 | - | 2 | 276 | |||
AK086063.1 | - | 2 | 277 | |||
AY046597.1 | - | 2 | 306 | |||
AY047360.1 | - | 2 | 251 | |||
BC027684.1 | - | 2 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK156237.1 | - | 496 | Chr. - - [-] on Build |