Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6030458C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029998.3 | NP_084274.1 | 4 | 846 | ||
GenBank mRNA | AK035859.1 | - | 4 | 831 | ||
AK036721.1 | - | 4 | 736 | |||
AK037867.1 | - | 3 | 498 | |||
AK038561.1 | - | 4 | 831 | |||
AK054133.1 | - | 4 | 820 | |||
AK081625.1 | - | 4 | 812 | |||
AK144748.1 | - | 4 | 801 | |||
AK152195.1 | - | 4 | 839 | |||
AK162187.1 | - | 4 | 863 | |||
AK163412.1 | - | 4 | 827 | |||
AK171821.1 | - | 3 | 674 | |||
BC119058.1 | - | 4 | 781 | 8733807 | ||
BC137759.1 | - | 4 | 781 | 8860778 |