Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6030458C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029998.3 | NP_084274.1 | 3, 4 | 489 | ||
GenBank mRNA | AK035859.1 | - | 3, 4 | 474 | ||
AK036721.1 | - | 3, 4 | 379 | |||
AK038561.1 | - | 3, 4 | 474 | |||
AK054133.1 | - | 3, 4 | 463 | |||
AK081625.1 | - | 3, 4 | 455 | |||
AK144748.1 | - | 3, 4 | 442 | |||
AK152195.1 | - | 3, 4 | 482 | |||
AK162187.1 | - | 3, 4 | 503 | |||
AK163412.1 | - | 3, 4 | 469 | |||
AK171821.1 | - | 2, 3 | 317 | |||
BC119058.1 | - | 3, 4 | 424 | 8733807 | ||
BC137759.1 | - | 3, 4 | 424 | 8860778 |