Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030138.2 | NP_084414.1 | 23 | 4974 | ||
GenBank mRNA | AK035342.1 | - | 1 | 1442 | ||
AK129041.1 | - | 23 | 4951 | |||
AK147319.1 | - | 22 | 4846 | |||
BC029317.1 | - | 1 | 1621 |