Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019470.1 | - | 10 | 3554 | ||
AK030347.1 | - | 10 | 2692 | |||
AK050075.1 | - | 10 | 2939 | |||
AK090330.1 | - | 8 | 2762 | |||
AK163883.1 | - | 10 | 3315 |