Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 14 | 1405 | ||
NM_001324172.1 | NP_001311101.1 | 14 | 1405 | |||
NM_030581.3 | NP_085058.3 | 14 | 1378 | |||
XM_005256146.3 | XP_005256203.1 | 14 | 1382 | |||
XM_011523331.2 | XP_011521633.1 | 12 | 1142 | |||
XM_011523332.2 | XP_011521634.1 | 8 | 830 | |||
XM_017023667.1 | XP_016879156.1 | 13 | 1239 | |||
XM_017023668.1 | XP_016879157.1 | 12 | 1015 | |||
XM_017023669.1 | XP_016879158.1 | 12 | 1030 | |||
XM_017023670.1 | XP_016879159.1 | 11 | 1063 | |||
GenBank mRNA | AB067510.1 | - | 9 | 851 | ||
AF370390.1 | - | 2 | 310 | |||
AK091316.1 | - | 2 | 158 | |||
AK302557.1 | - | 14 | 1446 | |||
BC004519.2 | - | 14 | 1342 | 3938206 | ||
BC014887.2 | - | 8 | 770 |