Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030581.3 | NP_085058.3 | 26 | 3366 | ||
XM_005256146.3 | XP_005256203.1 | 27 | 3427 | |||
XM_011523331.2 | XP_011521633.1 | 25 | 3187 | |||
XM_011523332.2 | XP_011521634.1 | 21 | 2875 | |||
XM_017023667.1 | XP_016879156.1 | 25 | 3227 | |||
XM_017023668.1 | XP_016879157.1 | 24 | 3003 | |||
XM_017023669.1 | XP_016879158.1 | 25 | 3075 | |||
XM_017023670.1 | XP_016879159.1 | 24 | 3108 | |||
GenBank mRNA | AB067510.1 | - | 21 | 2839 | ||
AF370390.1 | - | 14 | 2298 | |||
AK022332.1 | - | 9 | 1798 | |||
AK091316.1 | - | 14 | 2146 | |||
AL832078.1 | - | 11 | 1774 | |||
BC014887.2 | - | 20 | 2758 |