Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 3 | 310 | ||
NM_001324172.1 | NP_001311101.1 | 3 | 310 | |||
NM_030581.3 | NP_085058.3 | 3 | 283 | |||
XM_005256146.3 | XP_005256203.1 | 3 | 287 | |||
XM_011523331.2 | XP_011521633.1 | 2 | 127 | |||
XM_017023667.1 | XP_016879156.1 | 2 | 127 | |||
XM_017023670.1 | XP_016879159.1 | 1 | 23 | |||
GenBank mRNA | AK302557.1 | - | 3 | 351 | ||
BC004519.2 | - | 3 | 247 | 3938206 | ||
BU850134.1 | - | 3 | 235 | |||
CA777098.1 | - | 3 | 313 |