Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030842.1 | NP_110469.1 | 24, 25 | 3228 | ||
XM_006240800.2 | XP_006240862.1 | 24, 25 | 3418 | |||
XM_008765044.1 | XP_008763266.1 | 21, 22 | 2868 | |||
GenBank mRNA | BC088846.1 | - | 24, 25 | 3228 | 7313230 | |
X65036.1 | - | 24, 25 | 3098 |