Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032449.2 | NP_115825.1 | 24 | 3798 | ||
XM_005270590.2 | XP_005270647.1 | 25 | 3590 | |||
GenBank mRNA | AB058739.1 | - | 20 | 3229 | ||
AK095487.1 | - | 1 | 332 |