Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134760.1 | NP_001128232.1 | 11 | 1799 | ||
NM_032619.2 | NP_116008.2 | 12 | 1688 | |||
XM_006228691.2 | XP_006228753.1 | 11 | 1943 | |||
XM_006228692.2 | XP_006228754.1 | 12 | 1832 | |||
GenBank mRNA | AB260892.1 | - | 5 | 665 | ||
AF260436.1 | - | 3 | 352 | |||
BC086392.1 | - | 8 | 913 | |||
BC169048.1 | - | 12 | 1643 |