Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080453.2 | NP_536701.1 | 8 | 1395 | ||
NM_172797.2 | NP_766385.1 | 8 | 1353 | |||
GenBank mRNA | AK051258.1 | - | 8 | 1416 | ||
AY065653.1 | - | 8 | 1425 | |||
AY071828.1 | - | 8 | 1395 | |||
AY071829.1 | - | 8 | 1353 | |||
BC137921.1 | - | 8 | 1308 | 8860940 | ||
BC145048.1 | - | 8 | 1380 | 9053596 | ||
BC145049.1 | - | 8 | 1350 | 9053597 |