Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048054.2 | NP_001041519.1 | 4 | 1133 | ||
NM_130447.3 | NP_569714.2 | 4 | 1133 | |||
GenBank mRNA | AB052157.1 | - | 4 | 852 | ||
AF345951.1 | - | 4 | 977 | |||
AF345953.1 | - | 4 | 977 | |||
AK035652.1 | - | 4 | 879 | |||
AK144800.1 | - | 4 | 1134 | |||
BC057321.1 | - | 4 | 984 | 6830600 | ||
BC059232.1 | - | 4 | 920 | 6409944 | ||
CN529289.1 | - | 3 | 219 | |||
CV557369.1 | - | 3 | 409 |