Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133411.1 | NP_596902.1 | 16 | 2130 | ||
XM_008770939.1 | XP_008769161.1 | 16 | 2518 | |||
GenBank mRNA | AF534126.1 | - | 16 | 2141 | ||
AY533524.1 | - | 16 | 2130 | |||
AY533525.1 | - | 15 | 2019 |