Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 6, 7 | 971 | ||
NM_178089.2 | NP_835190.1 | 6, 7 | 952 | |||
GenBank mRNA | AK042090.1 | - | 6, 7 | 952 | ||
AK087545.1 | - | 6, 7 | 941 | |||
AK143666.1 | - | 6, 7 | 1014 | |||
AK155604.1 | - | 6, 7 | 982 | |||
AK183136.1 | - | 1, 2 | 44 | |||
AK192923.1 | - | 1, 2 | 44 | |||
AK195535.1 | - | 2, 3 | 203 | |||
BC021506.1 | - | 6, 7 | 995 | 5347099 | ||
BC027844.1 | - | 3, 4 | 264 | |||
BC078642.1 | - | 6, 7 | 895 | 6329630 |