Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290747.1 | NP_001277676.1 | 6 | 939 | ||
NM_145224.3 | NP_660259.1 | 9 | 1196 | |||
NM_181319.5 | NP_851836.2 | 9 | 1300 | |||
GenBank mRNA | AF516208.1 | - | 9 | 1300 | ||
AK053430.1 | - | 6 | 816 | |||
AK084312.1 | - | 9 | 1197 | |||
AY125891.1 | - | 8 | 1152 | |||
BC152402.1 | - | 6 | 939 | 40140811 |