Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 18 | 2958 | ||
XM_011513811.2 | XP_011512113.1 | 13 | 2142 | |||
XM_017007834.1 | XP_016863323.1 | 18 | 2484 | |||
GenBank mRNA | AK027494.1 | - | 7 | 1011 | ||
AK095866.1 | - | 13 | 2140 | |||
AK299168.1 | - | 15 | 2245 | |||
AL832257.1 | - | 10 | 1427 | |||
AY181243.1 | - | 16 | 2259 | |||
BC035645.1 | - | 18 | 2969 | |||
BX648287.1 | - | 2 | 2423 |
Set Membership: |
G-protein coupled receptor |