Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146518.1 | NP_666729.1 | 1 | 575 | ||
GenBank mRNA | BC119240.1 | - | 1 | 618 | 8733989 | |
BC120529.1 | - | 1 | 618 | 40129452 |