Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042410.1 | NP_001035869.1 | 3, 4 | 372 | ||
NM_018089.2 | NP_060559.2 | 3, 4 | 445 | |||
XM_005246663.2 | XP_005246720.1 | 3, 4 | 364 | |||
XM_011511392.2 | XP_011509694.1 | 3, 4 | 445 | |||
XM_017004420.1 | XP_016859909.1 | 3, 4 | 364 | |||
XM_017004421.1 | XP_016859910.1 | 3, 4 | 445 | |||
GenBank mRNA | AF364318.1 | - | 3, 4 | 299 | ||
AK001277.1 | - | 3, 4 | 382 | |||
AK023206.1 | - | 3, 4 | 372 | |||
AK095304.1 | - | 3, 4 | 309 | |||
AK098282.1 | - | 1, 2 | 1426 | |||
AK225275.1 | - | 3, 4 | 370 | |||
AK225309.1 | - | 3, 4 | 346 | |||
AK303077.1 | - | 2, 3 | 178 | |||
BC000238.1 | - | 3, 4 | 327 | 3351742 | ||
BC008948.2 | - | 3, 4 | 356 | 2959748 | ||
DA226194.1 | - | 3, 4 | 445 | |||
DQ412564.1 | - | 3, 4 | 382 |