Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145415.2 | NP_663390.2 | 12 | 2819 | ||
GenBank mRNA | AK041915.1 | - | 1 | 47 | ||
AK052355.1 | - | 12 | 2820 | |||
AK147251.1 | - | 12 | 2791 |