Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000250.1 | NP_000241.1 | 4 | 695 | ||
GenBank mRNA | BC130476.1 | - | 4 | 569 | 40146507 | |
J02694.1 | - | 4 | 666 | |||
M19507.1 | - | 4 | 695 | |||
S56200.1 | - | 4 | 518 | |||
X04876.1 | - | 4 | 681 |