Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021449.3 | NP_067424.2 | 11 | 1379 | ||
NM_175357.3 | NP_780566.1 | 11 | 1382 | |||
GenBank mRNA | AF229032.1 | - | 11 | 1334 | ||
AK032836.1 | - | 11 | 1356 | |||
AK050557.1 | - | 11 | 1365 | |||
AK075959.1 | - | 11 | 1355 | |||
AK076144.1 | - | 11 | 1362 | |||
AK077707.1 | - | 11 | 1362 | |||
AK160219.1 | - | 11 | 1358 | |||
BC033458.1 | - | 11 | 1340 | |||
BC046967.1 | - | 10 | 1297 | |||
BC069905.1 | - | 11 | 1339 | |||
BC086488.1 | - | 11 | 1357 | 30443656 |