Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027321.3 | NP_081597.1 | 3 | 369 | ||
NM_175413.3 | NP_780622.1 | 3 | 369 | |||
GenBank mRNA | AK008119.1 | - | 3 | 337 | ||
AK035072.1 | - | 3 | 371 | |||
AK039277.1 | - | 3 | 366 | |||
AK046671.1 | - | 3 | 265 | |||
AK052159.1 | - | 3 | 321 | |||
AK079125.1 | - | 3 | 347 | |||
BC119529.2 | - | 3 | 318 | 40056120 | ||
BC119530.1 | - | 3 | 319 | 40056121 |