Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175451.1 | NP_780660.1 | 2 | 2122 | ||
GenBank mRNA | AK030708.1 | - | 2 | 2007 | ||
AK089935.1 | - | 1 | 1369 | |||
BC138318.1 | - | 3 | 2144 | 8861338 | ||
BC138320.1 | - | 3 | 2144 | 8861340 |