Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307977.1 | NP_001294906.1 | 4 | 442 | ||
NM_018093.3 | NP_060563.2 | 5 | 521 | |||
XM_005274055.2 | XP_005274112.1 | 5 | 2382 | |||
GenBank mRNA | AK001301.1 | - | 5 | 521 | ||
AK292330.1 | - | 5 | 891 | |||
AK303186.1 | - | 4 | 363 | |||
AY500828.1 | - | 4 | 373 | |||
BC006351.2 | - | 4 | 364 | 4111690 |