Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181390.3 | NP_852055.1 | 3 | 731 | ||
GenBank mRNA | AJ277212.1 | - | 3 | 874 | ||
AK003360.1 | - | 3 | 447 | |||
AK003945.1 | - | 3 | 449 | |||
BC116939.1 | - | 3 | 439 | 40126258 | ||
BC116941.1 | - | 3 | 444 | 40126260 |