Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157904.1 | - | 1 | 417 | 9088095 | |
BC158074.1 | - | 1 | 417 | 9007371 | ||
BC172011.1 | - | 1 | 417 | 9054297 |
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053263.1 | NP_444493.1 | 4 | 448 | ||
NM_146130.3 | NP_666242.2 | 4 | 514 | |||
NM_198090.2 | NP_932758.1 | 4 | 514 | |||
GenBank mRNA | AB201711.1 | - | 4 | 494 | ||
AF463524.1 | - | 4 | 426 | |||
AK010481.1 | - | 4 | 442 | |||
AK133860.1 | - | 4 | 513 | |||
AK169663.1 | - | 4 | 496 | |||
BC023828.1 | - | 4 | 467 | 5343619 | ||
BC023908.1 | - | 4 | 483 | 5322033 | ||
BC024454.1 | - | 1 | 31 | |||
BC038364.1 | - | 4 | 496 | 5362605 | ||
BC057655.1 | - | 4 | 427 | |||
BC062198.1 | - | 4 | 505 | 2655879 | ||
BC064824.1 | - | 4 | 479 | 6477672 | ||
BC085474.1 | - | 3 | 389 | 30538435 | ||
BC092083.1 | - | 1 | 109 | |||
BC116250.1 | - | 4 | 417 | 40093600 | ||
BC116251.1 | - | 4 | 417 | 40093602 | ||
BC157919.1 | - | 4 | 417 | 9007347 | ||
BC158038.1 | - | 4 | 417 | 9007341 | ||
CO045618.1 | - | 3 | 298 |