Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028532.3 | NP_082808.1 | 7 | 516 | ||
NM_199455.2 | NP_955534.1 | 8 | 741 | |||
GenBank mRNA | AK006802.1 | - | 7 | 503 | ||
BC087870.1 | - | 8 | 737 | 6707505 | ||
BC104382.1 | - | 7 | 515 | |||
BC104383.1 | - | 7 | 516 | |||
BY706785.1 | - | 7 | 501 |