Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 3671 | ||
NM_001278662.1 | NP_001265591.1 | 5 | 3651 | |||
NM_001278663.1 | NP_001265592.1 | 3 | 3448 | |||
NM_001278664.1 | NP_001265593.1 | 3 | 3409 | |||
NM_001278677.1 | NP_001265606.1 | 6 | 3743 | |||
NM_001278678.1 | NP_001265607.1 | 2 | 3313 | |||
NM_203282.3 | NP_975011.3 | 4 | 3536 | |||
XM_011528443.1 | XP_011526745.1 | 6 | 3804 | |||
XM_011528444.1 | XP_011526746.1 | 7 | 3824 | |||
XM_011528448.2 | XP_011526750.1 | 4 | 3483 | |||
XM_017027514.1 | XP_016883003.1 | 6 | 3734 | |||
XM_017027515.1 | XP_016883004.1 | 8 | 4161 | |||
XM_017027516.1 | XP_016883005.1 | 4 | 6208 | |||
XM_017027517.1 | XP_016883006.1 | 5 | 4037 | |||
XM_017027518.1 | XP_016883007.1 | 4 | 3533 | |||
XM_017027519.1 | XP_016883008.1 | 5 | 3812 | |||
GenBank mRNA | AK303431.1 | - | 3 | 3382 | ||
AL832713.1 | - | 1 | 2232 | |||
BC043147.1 | - | 4 | 3505 | 5296940 |