Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011877.2 | NP_001011877.1 | 1 | 249 | ||
GenBank mRNA | BC141252.1 | - | 1 | 249 | 9055864 |