Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012476.2 | NP_001012494.1 | 2 | 631 | ||
XM_006251899.2 | XP_006251961.1 | 2 | 867 | |||
XM_008770562.1 | XP_008768784.1 | 2 | 639 | |||
GenBank mRNA | BC128714.1 | - | 2 | 631 | 7134188 |