Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017484.1 | NP_001017484.1 | 2, 3 | 111 | ||
XM_006248178.2 | XP_006248240.1 | 3, 4 | 337 | |||
XM_006248179.2 | XP_006248241.1 | 4, 5 | 439 | |||
XM_006248180.2 | XP_006248242.1 | 3, 4 | 386 | |||
XM_006248181.2 | XP_006248243.1 | 3, 4 | 277 | |||
XM_006248182.2 | XP_006248244.1 | 2, 3 | 175 | |||
GenBank mRNA | BC082082.1 | - | 2, 3 | 111 | 7133412 |