Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018008.3 | NP_060478.3 | 5 | 1787 | ||
GenBank mRNA | AF332890.1 | - | 4 | 1639 | ||
AK001004.1 | - | 3 | 1322 | |||
AK290464.1 | - | 5 | 1787 | |||
BC022464.1 | - | 5 | 1618 | 4794121 |