Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017486.1 | NP_001017486.1 | 2 | 154 | ||
XM_006249280.2 | XP_006249342.1 | 2 | 215 | |||
XM_008769201.1 | XP_008767423.1 | 2 | 216 | |||
GenBank mRNA | BC086385.1 | - | 2 | 154 | 7302515 |