Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025141.1 | NP_001020312.1 | 6 | 1483 | ||
XM_006251901.2 | XP_006251963.1 | 7 | 1893 | |||
XM_006251903.2 | XP_006251965.1 | 7 | 2189 | |||
XM_006251904.1 | XP_006251966.1 | 5 | 1370 | |||
XM_008770563.1 | XP_008768785.1 | 7 | 2344 | |||
XM_008770564.1 | XP_008768786.1 | 8 | 2241 | |||
XM_008770565.1 | XP_008768787.1 | 8 | 2114 | |||
XM_008770566.1 | XP_008768788.1 | 7 | 2175 | |||
XM_008770567.1 | XP_008768789.1 | 6 | 1996 | |||
XM_008770568.1 | XP_008768790.1 | 7 | 2752 | |||
GenBank mRNA | BC091423.1 | - | 6 | 1483 | 7365771 |