Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046234.1 | - | 2522 | 6816957 | ||
BC059048.1 | - | 2522 | 6816957 |