Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028148.2 | NP_082424.2 | 15 | 4570 | ||
GenBank mRNA | AK012092.1 | - | 5 | 3415 | ||
AK129478.1 | - | 16 | 4396 | |||
BC023842.1 | - | 5 | 1377 | |||
BC048359.1 | - | 5 | 883 | |||
BC055486.1 | - | 5 | 1242 | |||
BC058682.1 | - | 15 | 4549 |