Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sass6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289568.1 | NP_001276497.1 | 18 | 2220 | ||
NM_001289571.1 | NP_001276500.1 | 16 | 2076 | |||
NM_028349.3 | NP_082625.2 | 17 | 2137 | |||
GenBank mRNA | AK162883.1 | - | 3 | 1498 | ||
AK165224.1 | - | 18 | 2166 | |||
BC050110.1 | - | 17 | 2087 | 30024366 |