Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028428.2 | NP_082704.1 | 3 | 1530 | ||
GenBank mRNA | AJ542393.1 | - | 3 | 1529 | ||
AK029717.1 | - | 3 | 1530 | |||
AK030310.1 | - | 3 | 1529 | |||
AK032390.1 | - | 2 | 1612 | |||
AK034234.1 | - | 106 | ||||
AK046461.1 | - | 3 | 1530 | |||
AK172061.1 | - | 1 | 1533 | |||
BC138555.1 | - | 3 | 1529 | 8861575 | ||
BC145735.1 | - | 3 | 1529 | 40131038 |