Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017525.1 | NP_001017525.1 | 15 | 2757 | ||
NM_028709.2 | NP_082985.2 | 17 | 4610 | |||
GenBank mRNA | AK039038.1 | - | 14 | 2553 | ||
AK044022.1 | - | 15 | 2648 | |||
BC059036.1 | - | 5 | 1628 | |||
BC072592.1 | - | 17 | 3236 | |||
BC096535.1 | - | 17 | 4610 |