Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143888.2 | NP_001137360.1 | 11 | 1860 | ||
NM_001143889.2 | NP_001137361.1 | 10 | 1641 | |||
NM_001143890.2 | NP_001137362.1 | 9 | 1524 | |||
NM_001300958.1 | NP_001287887.1 | 10 | 3652 | |||
NM_018045.7 | NP_060515.3 | 11 | 1809 | |||
NR_125352.1 | 12 | 1918 | ||||
GenBank mRNA | AK001138.1 | - | 11 | 1779 | ||
AK021440.1 | - | 7 | 828 | |||
AK225717.1 | - | 11 | 1779 | |||
BC010573.1 | - | 11 | 1768 | |||
BC026322.1 | - | 11 | 1885 | 4794111 | ||
BC028381.2 | - | 11 | 1781 | 4812029 | ||
BQ721446.1 | - | 1 | 528 | |||
BX641056.1 | - | 9 | 3618 | |||
CN302967.1 | - | 1 | 270 |