Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028805.2 | NP_083081.2 | 20 | 3095 | ||
GenBank mRNA | AK031630.1 | - | 20 | 3007 | ||
AK136322.1 | - | 20 | 3016 |