Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931428F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166394.1 | NP_001159866.1 | 6 | 1386 | ||
NM_028888.2 | NP_083164.1 | 7 | 1783 | |||
GenBank mRNA | AK016481.1 | - | 6 | 1721 | ||
AK136929.1 | - | 6 | 1548 | |||
BC043046.1 | - | 6 | 1543 | 5703896 | ||
BC059045.1 | - | 6 | 1321 | 6816353 |