Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037221.2 | NP_001032298.1 | 3 | 1155 | ||
NM_001163433.1 | NP_001156905.1 | 2 | 635 | |||
NM_028966.3 | NP_083242.1 | 3 | 1155 | |||
GenBank mRNA | AK034323.1 | - | 2 | 749 | ||
AK133470.1 | - | 3 | 1134 | |||
AK134243.1 | - | 3 | 1153 | |||
AK161277.1 | - | 2 | 629 | |||
AK163751.1 | - | 2 | 1176 | |||
BC145827.1 | - | 3 | 1021 | 40131130 | ||
DQ278487.1 | - | 3 | 932 |