Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029022.1 | NP_083298.1 | 7 | 1080 | ||
GenBank mRNA | AK014701.1 | - | 7 | 1074 | ||
AK028703.1 | - | 7 | 1054 | |||
AK143725.1 | - | 7 | 1078 | |||
AK165936.1 | - | 7 | 1077 | |||
BC015296.1 | - | 6 | 1014 | |||
BC139390.1 | - | 7 | 1075 | 8862412 | ||
BC139391.1 | - | 7 | 1075 | 8862413 |